2-Amino-N-(3-hydroxybutyl)-3-methylbutanamide hydrochloride
Supplier: Matrix Scientific
052981-500MG
052981-2.5G
101901-812EA
364.62
USD
101901-812
101901-814
2-Amino-N-(3-hydroxybutyl)-3-methylbutanamide hydrochloride
2-Amino-N-(3-hydroxybutyl)-3-methylbutanamide hydrochloride
Hazard: Irritant
Formula:
C₉H₂₁ClN₂O₂ |
MDL Number:
MFCD13562496 CAS Number: 1246172-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...