3-Amino-N-(3-hydroxybutyl)propanamide hydrochloride
Supplier: Matrix Scientific
052980-500MG
052980-2.5G
101901-808EA
364.62
USD
101901-808
101901-810
3-Amino-N-(3-hydroxybutyl)propanamide hydrochloride
3-Amino-N-(3-hydroxybutyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₇ClN₂O₂ |
MDL Number:
MFCD13562495 CAS Number: 1220034-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...