N-(3-Hydroxybutyl)-2-(methylamino)acetamide hydrochloride
Supplier: Matrix Scientific
052979-500MG
052979-2.5G
101901-804EA
364.62
USD
101901-804
101901-806
N-(3-Hydroxybutyl)-2-(methylamino)acetamide hydrochloride
N-(3-Hydroxybutyl)-2-(methylamino)acetamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₇ClN₂O₂ |
MDL Number:
MFCD13562494 CAS Number: 1220037-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...