2-Amino-N-(3-hydroxybutyl)propanamide hydrochloride

Supplier: Matrix Scientific

052978-500MG 052978-2.5G
101901-800EA 364.62 USD
101901-800 101901-802
2-Amino-N-(3-hydroxybutyl)propanamide hydrochloride
2-Amino-N-(3-hydroxybutyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O₂
MDL Number: MFCD13562493
CAS Number: 1246172-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR