2-Amino-N-(3-hydroxybutyl)acetamide hydrochloride
Supplier: Matrix Scientific
052977-500MG
052977-2.5G
101901-796EA
364.62
USD
101901-796
101901-798
2-Amino-N-(3-hydroxybutyl)acetamide hydrochloride
2-Amino-N-(3-hydroxybutyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₆H₁₅ClN₂O₂ |
MDL Number:
MFCD13562492 CAS Number: 1220037-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...