4-[(6-Chloro-2-pyridinyl)amino]-2-butanol

Supplier: Matrix Scientific

052976-500MG 052976-2.5G
101901-792EA 364.62 USD
101901-792 101901-794
4-[(6-Chloro-2-pyridinyl)amino]-2-butanol
4-[(6-Chloro-2-pyridinyl)amino]-2-butanol

Hazard: Irritant

Formula: C₉H₁₃ClN₂O
MDL Number: MFCD13562491
CAS Number: 1220028-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR