4-[(5-Bromo-2-pyridinyl)amino]-2-butanol
Supplier: Matrix Scientific
052975-500MG
052975-2.5G
101901-788EA
364.62
USD
101901-788
101901-790
4-[(5-Bromo-2-pyridinyl)amino]-2-butanol
4-[(5-Bromo-2-pyridinyl)amino]-2-butanol
Hazard: Irritant
Formula:
C₉H₁₃BrN₂O |
MDL Number:
MFCD13562490 CAS Number: 1220037-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...