4-[(3-Amino-2-pyridinyl)amino]-2-butanol

Supplier: Matrix Scientific

052972-500MG 052972-2.5G
101901-776EA 364.62 USD
101901-776 101901-778
4-[(3-Amino-2-pyridinyl)amino]-2-butanol
4-[(3-Amino-2-pyridinyl)amino]-2-butanol

Hazard: Irritant

Formula: C₉H₁₅N₃O
MDL Number: MFCD13562487
CAS Number: 1220034-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR