2-[(3-Hydroxybutyl)amino]nicotinic acid

Supplier: Matrix Scientific

052971-500MG 052971-2.5G
101901-772EA 364.62 USD
101901-772 101901-774
2-[(3-Hydroxybutyl)amino]nicotinic acid
2-[(3-Hydroxybutyl)amino]nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₃
MDL Number: MFCD13562486
CAS Number: 1220037-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR