2-[(3-Hydroxybutyl)amino]isonicotinic acid

Supplier: Matrix Scientific

052970-500MG 052970-2.5G
101901-768EA 364.62 USD
101901-768 101901-770
2-[(3-Hydroxybutyl)amino]isonicotinic acid
2-[(3-Hydroxybutyl)amino]isonicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₃
MDL Number: MFCD13562485
CAS Number: 1220035-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR