4-[(6-Chloro-4-pyrimidinyl)amino]-2-butanol

Supplier: Matrix Scientific

052967-500MG 052967-2.5G
101901-756EA 364.62 USD
101901-756 101901-758
4-[(6-Chloro-4-pyrimidinyl)amino]-2-butanol
4-[(6-Chloro-4-pyrimidinyl)amino]-2-butanol

Hazard: Irritant

Formula: C₈H₁₂ClN₃O
MDL Number: MFCD13562482
CAS Number: 1219982-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR