4-[(5-Bromo-4-methyl-2-pyridinyl)amino]-2-butanol
Supplier: Matrix Scientific
052966-500MG
052966-2.5G
101901-752EA
364.62
USD
101901-752
101901-754
4-[(5-Bromo-4-methyl-2-pyridinyl)amino]-2-butanol
4-[(5-Bromo-4-methyl-2-pyridinyl)amino]-2-butanol
Hazard: Irritant
Formula:
C₁₀H₁₅BrN₂O |
MDL Number:
MFCD13562481 CAS Number: 1220030-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...