4-[(3-Hydroxybutyl)amino]-3-nitrobenzoic acid

Supplier: Matrix Scientific

052965-500MG 052965-2.5G
101901-748EA 364.62 USD
101901-748 101901-750
4-[(3-Hydroxybutyl)amino]-3-nitrobenzoic acid
4-[(3-Hydroxybutyl)amino]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₅
MDL Number: MFCD13562480
CAS Number: 1219957-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR