Ethyl-3-amino-4-[(3-hydroxybutyl)amino]benzoate

Supplier: Matrix Scientific

052964-500MG 052964-2.5G
101901-744EA 364.62 USD
101901-744 101901-746
Ethyl-3-amino-4-[(3-hydroxybutyl)amino]benzoate
Ethyl-3-amino-4-[(3-hydroxybutyl)amino]benzoate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₃
MDL Number: MFCD13562479
CAS Number: 1220018-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR