N-Isobutyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052961-500MG 052961-2.5G
101901-732EA 364.62 USD
101901-732 101901-734
N-Isobutyl-4-piperidinecarboxamide hydrochloride
N-Isobutyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562477
CAS Number: 1220037-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR