2-Amino-N-isobutyl-2-methylpropanamide hydrochloride
Supplier: Matrix Scientific
052956-500MG
052956-2.5G
101901-712EA
364.62
USD
101901-712
101901-714
2-Amino-N-isobutyl-2-methylpropanamide hydrochloride
2-Amino-N-isobutyl-2-methylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₉ClN₂O |
MDL Number:
MFCD13562473 CAS Number: 1220035-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...