2-Amino-N-isobutyl-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052955-500MG 052955-2.5G
101901-708EA 364.62 USD
101901-708 101901-710
2-Amino-N-isobutyl-3-methylbutanamide hydrochloride
2-Amino-N-isobutyl-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₉H₂₁ClN₂O
MDL Number: MFCD13562472
CAS Number: 1236255-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR