2-Amino-N-isobutylpropanamide hydrochloride

Supplier: Matrix Scientific

052952-500MG 052952-2.5G
101901-696EA 364.62 USD
101901-696 101901-698
2-Amino-N-isobutylpropanamide hydrochloride
2-Amino-N-isobutylpropanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MDL Number: MFCD13562469
CAS Number: 1236262-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR