5-Bromo-N-isobutyl-2-pyridinamine

Supplier: Matrix Scientific

052949-500MG 052949-2.5G
101901-684EA 364.62 USD
101901-684 101901-686
5-Bromo-N-isobutyl-2-pyridinamine
5-Bromo-N-isobutyl-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃BrN₂
MDL Number: MFCD00956963
CAS Number: 300394-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR