5-Bromo-N-isobutyl-3-methyl-2-pyridinamine

Supplier: Matrix Scientific

052948-500MG 052948-2.5G
101901-680EA 364.62 USD
101901-680 101901-682
5-Bromo-N-isobutyl-3-methyl-2-pyridinamine
5-Bromo-N-isobutyl-3-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂
MDL Number: MFCD13562466
CAS Number: 1220037-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR