2-(Isobutylamino)nicotinic acid

Supplier: Matrix Scientific

052945-500MG 052945-2.5G
101901-668EA 112.31 USD
101901-668 101901-670
2-(Isobutylamino)nicotinic acid
2-(Isobutylamino)nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD11106740
CAS Number: 1019461-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR