6-Chloro-N-isobutyl-4-pyrimidinamine

Supplier: Matrix Scientific

052941-500MG 052941-2.5G
101901-652EA 364.62 USD
101901-652 101901-654
6-Chloro-N-isobutyl-4-pyrimidinamine
6-Chloro-N-isobutyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₈H₁₂ClN₃
MDL Number: MFCD13562464
CAS Number: 1220028-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR