5-Bromo-N-isobutyl-4-methyl-2-pyridinamine

Supplier: Matrix Scientific

052940-500MG 052940-2.5G
101901-648EA 364.62 USD
101901-648 101901-650
5-Bromo-N-isobutyl-4-methyl-2-pyridinamine
5-Bromo-N-isobutyl-4-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂
MW: 243.15 g/mol
MDL Number: MFCD13562463
CAS Number: 1220028-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR