N-(sec-Butyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052933-500MG 052933-2.5G
101901-620EA 364.62 USD
101901-620 101901-622
N-(sec-Butyl)-2-piperidinecarboxamide hydrochloride
N-(sec-Butyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562459
CAS Number: 1246172-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR