2-Amino-N-(sec-butyl)-2-methylpropanamide hydrochloride

Supplier: Matrix Scientific

052930-500MG 052930-2.5G
101901-608EA 364.62 USD
101901-608 101901-610
2-Amino-N-(sec-butyl)-2-methylpropanamide hydrochloride
2-Amino-N-(sec-butyl)-2-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₉ClN₂O
MDL Number: MFCD13562456
CAS Number: 1220029-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR