2-Amino-N-(sec-butyl)-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052929-500MG 052929-2.5G
101901-604EA 364.62 USD
101901-604 101901-606
2-Amino-N-(sec-butyl)-3-methylbutanamide hydrochloride
2-Amino-N-(sec-butyl)-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₉H₂₁ClN₂O
MDL Number: MFCD13562455
CAS Number: 1236262-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR