3-Amino-N-(sec-butyl)propanamide hydrochloride

Supplier: Matrix Scientific

052928-500MG 052928-2.5G
101901-600EA 364.62 USD
101901-600 101901-602
3-Amino-N-(sec-butyl)propanamide hydrochloride
3-Amino-N-(sec-butyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MDL Number: MFCD12197107
CAS Number: 1181458-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR