N-(sec-Butyl)-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052927-500MG 052927-2.5G
101901-596EA 364.62 USD
101901-596 101901-598
N-(sec-Butyl)-2-(methylamino)acetamide hydrochloride
N-(sec-Butyl)-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MDL Number: MFCD13562454
CAS Number: 1219957-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR