2-Amino-N-(sec-butyl)acetamide hydrochloride
Supplier: Matrix Scientific
052925-500MG
052925-2.5G
101901-588EA
364.62
USD
101901-588
101901-590
2-Amino-N-(sec-butyl)acetamide hydrochloride
2-Amino-N-(sec-butyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₆H₁₅ClN₂O |
MDL Number:
MFCD13562452 CAS Number: 1220019-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...