N-(sec-Butyl)-6-chloro-2-pyridinamine

Supplier: Matrix Scientific

052924-500MG 052924-2.5G
101901-584EA 364.62 USD
101901-584 101901-586
N-(sec-Butyl)-6-chloro-2-pyridinamine
N-(sec-Butyl)-6-chloro-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃ClN₂
MDL Number: MFCD13562451
CAS Number: 1220029-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR