N-(sec-Butyl)-2-chloronicotinamide

Supplier: Matrix Scientific

052921-500MG 052921-2.5G
101901-572EA 364.62 USD
101901-572 101901-574
N-(sec-Butyl)-2-chloronicotinamide
N-(sec-Butyl)-2-chloronicotinamide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD03396426
CAS Number: 546116-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR