N2-(sec-Butyl)-2,3-pyridinediamine

Supplier: Matrix Scientific

052920-500MG 052920-2.5G
101901-568EA 364.62 USD
101901-568 101901-570
N2-(sec-Butyl)-2,3-pyridinediamine
N2-(sec-Butyl)-2,3-pyridinediamine

Hazard: Irritant

Formula: C₉H₁₅N₃
MDL Number: MFCD11186917
CAS Number: 1040042-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR