2-(sec-Butylamino)nicotinic acid

Supplier: Matrix Scientific

052919-500MG 052919-2.5G
101901-564EA 364.62 USD
101901-564 101901-566
2-(sec-Butylamino)nicotinic acid
2-(sec-Butylamino)nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD11132001
CAS Number: 1019388-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR