2-(sec-Butylamino)isonicotinic acid

Supplier: Matrix Scientific

052918-500MG 052918-2.5G
101901-560EA 364.62 USD
101901-560 101901-562
2-(sec-Butylamino)isonicotinic acid
2-(sec-Butylamino)isonicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MW: 194.24 g/mol
MDL Number: MFCD11131996
CAS Number: 1019388-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR