N-(5-Bromo-4-methyl-2-pyridinyl)-N-(sec-butyl)amine

Supplier: Matrix Scientific

052914-500MG 052914-2.5G
101901-544EA 364.62 USD
101901-544 101901-546
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(sec-butyl)amine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(sec-butyl)amine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂
MDL Number: MFCD13562447
CAS Number: 1220027-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR