4-(sec-Butylamino)-3-nitrobenzoic acid

Supplier: Matrix Scientific

052913-500MG 052913-2.5G
101901-540EA 364.62 USD
101901-540 101901-542
4-(sec-Butylamino)-3-nitrobenzoic acid
4-(sec-Butylamino)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₄
MDL Number: MFCD02937559
CAS Number: 440347-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR