N-(tert-Butyl)-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052908-500MG 052908-2.5G
101901-520EA 364.62 USD
101901-520 101901-522
N-(tert-Butyl)-3-piperidinecarboxamide hydrochloride
N-(tert-Butyl)-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562445
CAS Number: 937725-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR