N-(tert-Butyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

052907-500MG 052907-2.5G
101901-516EA 364.62 USD
101901-516 101901-518
N-(tert-Butyl)-2-piperidinecarboxamide hydrochloride
N-(tert-Butyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562444
CAS Number: 1236261-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR