2-Amino-N-(tert-butyl)-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

052905-500MG 052905-2.5G
101901-508EA 149.23 USD
101901-508 101901-510
2-Amino-N-(tert-butyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(tert-butyl)-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁ClN₂O
MDL Number: MFCD13562442
CAS Number: 1236262-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR