2-Amino-N-(tert-butyl)-2-methylpropanamide hydrochloride
Supplier: Matrix Scientific
052904-500MG
052904-2.5G
101901-504EA
364.62
USD
101901-504
101901-506
2-Amino-N-(tert-butyl)-2-methylpropanamide hydrochloride
2-Amino-N-(tert-butyl)-2-methylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₉ClN₂O |
MDL Number:
MFCD13562441 CAS Number: 1220037-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...