3-Amino-N-(tert-butyl)propanamide hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-Amino-N-(tert-butyl)propanamide

052902-500MG 052902-2.5G
101901-496EA 364.62 USD
101901-496 101901-498
3-Amino-N-(tert-butyl)propanamide hydrochloride
3-Amino-N-(tert-butyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MW: 180.68 g/mol
MDL Number: MFCD11109909
CAS Number: 209467-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR