N-(tert-Butyl)-2-(methylamino)acetamide hydrochloride
Supplier: Matrix Scientific
052901-500MG
052901-2.5G
101901-492EA
384.62
USD
101901-492
101901-494
N-(tert-Butyl)-2-(methylamino)acetamide hydrochloride
N-(tert-Butyl)-2-(methylamino)acetamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₇ClN₂O |
MDL Number:
MFCD13562439 CAS Number: 1220037-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...