2-Amino-N-(tert-butyl)propanamide hydrochloride

Supplier: Matrix Scientific

052900-500MG 052900-2.5G
101901-488EA 364.62 USD
101901-488 101901-490
2-Amino-N-(tert-butyl)propanamide hydrochloride
2-Amino-N-(tert-butyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O
MDL Number: MFCD13196116
CAS Number: 1214031-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR