N-(tert-Butyl)-6-chloro-2-pyridinamine

Supplier: Matrix Scientific

052899-500MG 052899-2.5G
101901-484EA 364.62 USD
101901-484 101901-486
N-(tert-Butyl)-6-chloro-2-pyridinamine
N-(tert-Butyl)-6-chloro-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃ClN₂
MDL Number: MFCD13562438
CAS Number: 791095-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR