N-(5-Bromo-2-pyridinyl)-N-(tert-butyl)amine

Supplier: Matrix Scientific

052898-500MG 052898-2.5G
101901-480EA 364.62 USD
101901-480 101901-482
N-(5-Bromo-2-pyridinyl)-N-(tert-butyl)amine
N-(5-Bromo-2-pyridinyl)-N-(tert-butyl)amine

Hazard: Irritant

Formula: C₉H₁₃BrN₂
MDL Number: MFCD13562437
CAS Number: 1220034-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR