N-(5-Bromo-3-methyl-2-pyridinyl)-N-(tert-butyl)amine

Supplier: Matrix Scientific

052897-500MG 052897-2.5G
101901-476EA 364.62 USD
101901-476 101901-478
N-(5-Bromo-3-methyl-2-pyridinyl)-N-(tert-butyl)amine
N-(5-Bromo-3-methyl-2-pyridinyl)-N-(tert-butyl)amine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂
MDL Number: MFCD13562436
CAS Number: 1220018-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR