N-(tert-Butyl)-2-chloronicotinamide

Supplier: Matrix Scientific

052896-500MG 052896-2.5G
101901-472EA 364.62 USD
101901-472 101901-474
N-(tert-Butyl)-2-chloronicotinamide
N-(tert-Butyl)-2-chloronicotinamide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MW: 212.68 g/mol
MDL Number: MFCD00667768
CAS Number: 144084-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR