N2-(tert-Butyl)-2,3-pyridinediamine
Supplier: Matrix Scientific
052895-500MG
052895-2.5G
101901-468EA
364.62
USD
101901-468
101901-470
N2-(tert-Butyl)-2,3-pyridinediamine
N2-(tert-Butyl)-2,3-pyridinediamine
Hazard: Irritant
Formula:
C₉H₁₅N₃ |
MDL Number:
MFCD10688728 CAS Number: 1022146-68-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...