2-(tert-Butylamino)nicotinic acid

Supplier: Matrix Scientific

052894-500MG 052894-2.5G
101901-464EA 166.15 USD
101901-464 101901-466
2-(tert-Butylamino)nicotinic acid
2-(tert-Butylamino)nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD03119363
CAS Number: 460044-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR