N-(tert-Butyl)-3-chloro-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

052891-500MG 052891-2.5G
101901-452EA 364.62 USD
101901-452 101901-454
N-(tert-Butyl)-3-chloro-5-(trifluoromethyl)-2-pyridinamine
N-(tert-Butyl)-3-chloro-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₂ClF₃N₂
MDL Number: MFCD13562434
CAS Number: 1220037-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR